Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gys1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030678.3 | NP_109603.2 | 14 | 1895 | ||
GenBank mRNA | AK050813.1 | - | 12 | 1328 | ||
AK085476.1 | - | 10 | 923 | |||
AK169344.1 | - | 14 | 1891 | |||
AK170404.1 | - | 14 | 1897 | |||
AK170585.1 | - | 14 | 1815 | |||
AK171148.1 | - | 14 | 1850 | |||
AK171426.1 | - | 14 | 1874 | |||
BC019389.1 | - | 14 | 1688 | 3663506 | ||
BC131687.1 | - | 14 | 1742 | 40111722 | ||
BC131688.1 | - | 14 | 1742 | 40111723 | ||
BC152550.1 | - | 14 | 1742 | 40111724 | ||
U53218.1 | - | 14 | 1733 | |||
X94616.1 | - | 14 | 1742 |