Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-M9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008205.1 | NP_032231.1 | 5 | 931 | ||
GenBank mRNA | AF469488.1 | - | 5 | 931 | ||
AF539595.1 | - | 5 | 931 | |||
AK080256.1 | - | 4 | 679 | |||
BC106996.2 | - | 5 | 930 |