Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HK2 SIRNA Assays ›
Gene Symbol:
Hk2
Gene Name:
hexokinase 2
Gene Aliases:
AI642394, HKII
Chromosome Location:
Chr.6: 82725025 - 82774454 on Build GRCm38
UniGene ID:
Mm.255848
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS274840
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Contact Us
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hk2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_013820.3
NP_038848.1
17
2912
GenBank mRNA
AJ238540.1
-
17
2855
AK145936.1
-
17
2893
BC054472.1
-
17
2831
6465367
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
cellular glucose homeostasis
response to ischemia
carbohydrate metabolic process
glucose metabolic process
glycolytic process
metabolic process
apoptotic mitochondrial changes
phosphorylation
regulation of glucose import
carbohydrate phosphorylation
glucose 6-phosphate metabolic process
nucleotide binding
catalytic activity
glucokinase activity
hexokinase activity
ATP binding
glucose binding
fructokinase activity
kinase activity
transferase activity
phosphotransferase activity, alcohol group as acceptor
mannokinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0