Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025903.2 | NP_080179.1 | 2 | 274 | ||
GenBank mRNA | AK007687.1 | - | 2 | 275 | ||
AK149496.1 | - | 2 | 255 | |||
AK156625.1 | - | 2 | 268 | |||
AK165967.1 | - | 2 | 267 | |||
AK166130.1 | - | 2 | 342 | |||
AK166258.1 | - | 2 | 232 | |||
AY298860.1 | - | 2 | 103 | |||
BC023486.1 | - | 2 | 222 | |||
BC037434.1 | - | 2 | 230 | 2651172 |