Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inppl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122739.1 | NP_001116211.1 | 16, 17 | 2165 | ||
NM_010567.2 | NP_034697.2 | 16, 17 | 2399 | |||
GenBank mRNA | AF162781.1 | - | 16, 17 | 2249 | ||
AK133359.1 | - | 16, 17 | 2126 | |||
AK134307.1 | - | 1, 2 | 121 | |||
AK149873.1 | - | 16, 17 | 2183 | |||
BC039207.1 | - | 3, 4 | 301 | |||
BC063080.1 | - | 16, 17 | 2095 | 6416946 |