Search
Search

Lats1| Gene Symbol: Lats1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010690.1 | NP_034820.1 | 4 | 1853 | ||
| GenBank mRNA | AF104414.1 | - | 1 | 978 | ||
| AK038612.1 | - | 4 | 1854 | |||
| BC158092.1 | - | 3 | 1699 | 9088177 | ||
| BC158123.1 | - | 4 | 1848 | 9088196 | ||