Search
Search

Blnk| Gene Symbol: Blnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008528.4 | NP_032554.2 | 2 | 572 | ||
| GenBank mRNA | AB015290.1 | - | 2 | 246 | ||
| AF068182.1 | - | 2 | 227 | |||
| AJ222814.1 | - | 2 | 197 | |||
| AK089704.1 | - | 2 | 212 | |||
| AK172016.1 | - | 2 | 199 | |||
| BC059785.1 | - | 2 | 198 | 5339723 | ||
| Y17159.1 | - | 2 | 572 | |||