Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130879.2 | NP_570949.2 | 4 | 649 | ||
GenBank mRNA | AK052297.1 | - | 4 | 590 | ||
AK133251.1 | - | 5 | 666 | |||
AK166158.1 | - | 4 | 652 | |||
AK220190.1 | - | 4 | 610 | |||
BC021769.1 | - | 4 | 608 | |||
BC145295.1 | - | 5 | 640 |