siRNA Details
Gene Symbol: Mknk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB164081.1 | - | 11 | 980 | ||
AK008277.1 | - | 2 | 215 | |||
AK030830.1 | - | 6 | 532 | |||
AK154235.1 | - | 12 | 1260 | |||
AK164278.1 | - | 2 | 88 | |||
AK207448.1 | - | 2 | 182 | |||
BC010256.1 | - | 10 | 890 | 3257646 | ||
Y11092.1 | - | 10 | 931 |
