Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gadd45b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008655.1 | NP_032681.1 | 3 | 487 | ||
GenBank mRNA | AK010420.1 | - | 3 | 505 | ||
AK149895.1 | - | 3 | 505 | |||
AK154952.1 | - | 3 | 505 | |||
BC023815.1 | - | 3 | 499 | 5355150 | ||
X54149.1 | - | 3 | 487 |