Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other P4HB SIRNA Assays ›
          Gene Symbol:
          P4hb
          Gene Name:
          prolyl 4-hydroxylase, beta polypeptide
          Gene Aliases:
          ERp59, PDI, Pdia1, Thbp
          Chromosome Location:
          Chr.11: 120560304 - 120572936 on Build GRCm38
          UniGene ID:
          Mm.16660
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS276205
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: P4hb
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011032.2 NP_035162.1 5 842
          GenBank mRNA AK141304.1 - 5 779
          AK146542.1 - 5 780
          AK149605.1 - 5 781
          AK149965.1 - 5 780
          AK150002.1 - 5 806
          AK150422.1 - 5 780
          AK150592.1 - 5 780
          AK150755.1 - 5 766
          AK150805.1 - 5 780
          AK151351.1 - 5 774
          AK151546.1 - 5 780
          AK152141.1 - 5 780
          AK152217.1 - 5 780
          AK152791.1 - 5 780
          AK152834.1 - 5 780
          AK153365.1 - 5 780
          AK159606.1 - 5 780
          AK159729.1 - 5 780
          AK159965.1 - 5 780
          AK161537.1 - 5 780
          AK167796.1 - 5 780
          AK168330.1 - 5 778
          AK168336.1 - 5 762
          AK168612.1 - 5 780
          AK168893.1 - 5 780
          AK169265.1 - 5 777
          AK170603.1 - 5 777
          AK183040.1 - 1 71
          AK199187.1 - 1 71
          AK200312.1 - 1 71
          AK214426.1 - 2 148
          BC008549.1 - 5 768 3591404
          BC093512.1 - 5 759 5124376
          J05185.1 - 5 842
          X06453.1 - 5 782

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_012998, M21476, BC061857, X02918, M21018.

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          peptidyl-proline hydroxylation to 4-hydroxy-L-proline
          response to endoplasmic reticulum stress
          cell redox homeostasis
          cellular response to hypoxia
          regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
          protein disulfide isomerase activity
          procollagen-proline 4-dioxygenase activity
          isomerase activity
          enzyme binding
          peptidyl-proline 4-dioxygenase activity
          poly(A) RNA binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0