Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161406.1 | NP_001154878.1 | 4 | 803 | ||
NM_145890.2 | NP_665897.2 | 2 | 435 | |||
GenBank mRNA | AF411211.1 | - | 2 | 435 | ||
AF411212.1 | - | 4 | 774 | |||
AK076307.1 | - | 4 | 789 | |||
AK161048.1 | - | 4 | 685 | |||
AK167035.1 | - | 4 | 803 | |||
BC013080.1 | - | 4 | 743 | 3499860 | ||
BC037233.1 | - | 4 | 795 | 4951512 |