Search
Search

Six3| Gene Symbol: Six3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011381.4 | NP_035511.2 | 1 | 841 | ||
| GenBank mRNA | BC094426.1 | - | 644 | 6855602 | ||
| BC098096.1 | - | 1 | 461 | 6402835 | ||
| BF467709.1 | - | 1 | 479 | |||
| D83144.1 | - | 1 | 1102 | |||
| D83145.1 | - | 1 | 427 | |||
| X90871.1 | - | 616 | ||||