Search
Search

Slc7a7| Gene Symbol: Slc7a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253679.1 | NP_001240608.1 | 9 | 1427 | ||
| NM_001253680.1 | NP_001240609.1 | 8 | 1445 | |||
| NM_011405.4 | NP_035535.2 | 10 | 1584 | |||
| GenBank mRNA | AJ012754.1 | - | 8 | 1435 | ||
| AJ130943.1 | - | 9 | 1427 | |||
| AK150950.1 | - | 10 | 1584 | |||
| AK151718.1 | - | 10 | 1584 | |||
| AK152637.1 | - | 8 | 1350 | |||
| AK170851.1 | - | 8 | 1356 | |||
| BC014709.1 | - | 8 | 1433 | 4482816 | ||