siRNA Details
Gene Symbol: Sox18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009236.2 | NP_033262.2 | 2 | 949 | ||
GenBank mRNA | AK144281.1 | - | 2 | 950 | ||
BC006612.1 | - | 2 | 949 | 3967084 |

Gene Symbol: Sox18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009236.2 | NP_033262.2 | 2 | 949 | ||
GenBank mRNA | AK144281.1 | - | 2 | 950 | ||
BC006612.1 | - | 2 | 949 | 3967084 |