Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SOX2 SIRNA Assays ›
          Gene Symbol:
          Sox2
          Gene Name:
          SRY (sex determining region Y)-box 2
          Gene Aliases:
          Sox-2, lcc, ysb
          Chromosome Location:
          Chr.3: 34650005 - 34652461 on Build GRCm38
          UniGene ID:
          Mm.65396
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS277201
          Size
          Availability Made To Order
          Catalog #
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Sox2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011443.3 NP_035573.3 1 933
          GenBank mRNA AB108673.1 - 1 555
          AB221649.1 - 1 522
          AK131933.1 - 1 371
          AK149358.1 - 1 256
          BC057574.1 - 1 804 6413283
          U31967.1 - 1 883

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001109181.

          Panther Classification:

          Molecular Function -

          HMG box transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          osteoblast differentiation
          cell fate specification
          endodermal cell fate specification
          positive regulation of neuroblast proliferation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          cell cycle arrest
          multicellular organism development
          sensory perception of sound
          response to organic substance
          regulation of gene expression
          stem cell population maintenance
          forebrain neuron differentiation
          adenohypophysis development
          cerebral cortex development
          positive regulation of cell-cell adhesion
          negative regulation of Wnt signaling pathway
          male genitalia development
          positive regulation of epithelial cell differentiation
          olfactory placode formation
          response to retinoic acid
          somatic stem cell population maintenance
          inner ear morphogenesis
          regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of MAPK cascade
          tongue development
          cell fate commitment
          negative regulation of cell differentiation
          positive regulation of cell differentiation
          negative regulation of neuron differentiation
          positive regulation of neuron differentiation
          negative regulation of osteoblast differentiation
          positive regulation of Notch signaling pathway
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          pigment biosynthetic process
          lung alveolus development
          embryonic organ development
          anatomical structure formation involved in morphogenesis
          neuron fate commitment
          diencephalon morphogenesis
          stem cell differentiation
          negative regulation of epithelial cell proliferation
          regulation of neurogenesis
          detection of mechanical stimulus involved in sensory perception of sound
          detection of mechanical stimulus involved in equilibrioception
          retina morphogenesis in camera-type eye
          lens induction in camera-type eye
          epithelial tube branching involved in lung morphogenesis
          response to growth factor
          negative regulation of canonical Wnt signaling pathway
          neuronal stem cell population maintenance
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          chromatin DNA binding
          miRNA binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-kkgv7:80/100.66.73.110:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0