Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011446.1 | NP_035576.1 | 2 | 1126 | ||
GenBank mRNA | AB023419.1 | - | 2 | 1126 | ||
AK155670.1 | - | 2 | 1159 | |||
AK156497.1 | - | 2 | 1160 | |||
BC131936.1 | - | 2 | 1108 | 40130213 | ||
BC145925.1 | - | 2 | 1108 | 40131228 |