Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009282.3 | NP_033308.2 | 24 | 2691 | ||
GenBank mRNA | BC062954.1 | - | 24 | 2691 | 30537388 | |
BC069841.1 | - | 25 | 2619 | |||
Z75332.1 | - | 24 | 2740 |