Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stra13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016665.2 | NP_057874.2 | 2 | 90 | ||
GenBank mRNA | AK076181.1 | - | 2 | 62 | ||
AK080500.1 | - | 2 | 62 | |||
AK170126.1 | - | 2 | 62 | |||
BC052335.1 | - | 3 | 340 | |||
CA320983.1 | - | 1 | 29 | |||
CJ236047.1 | - | 2 | 91 | |||
U95003.1 | - | 3 | 154 | |||
U95005.1 | - | 3 | 328 |