Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Synj2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113351.1 | NP_001106822.1 | 14 | 2115 | ||
NM_001113352.2 | NP_001106823.1 | 15 | 2204 | |||
NM_001113353.2 | NP_001106824.1 | 15 | 2204 | |||
NM_001290698.1 | NP_001277627.1 | 9 | 1232 | |||
NM_011523.2 | NP_035653.2 | 14 | 2115 | |||
GenBank mRNA | AF041857.1 | - | 15 | 2205 | ||
AF041858.1 | - | 4 | 467 | |||
AF041859.1 | - | 8 | 996 | |||
AF041860.1 | - | 1 | 47 | |||
AF041862.1 | - | 14 | 2060 | |||
AK014584.1 | - | 11 | 1359 | |||
AK019694.1 | - | 9 | 1233 | |||
AK029755.1 | - | 12 | 1622 | |||
AK038038.1 | - | 12 | 1528 | |||
AK129122.1 | - | 13 | 1632 | |||
AK142919.1 | - | 14 | 1836 | |||
AK148053.1 | - | 15 | 1942 | |||
AK161467.1 | - | 9 | 1216 | |||
AK163512.1 | - | 13 | 1845 | |||
BC058749.1 | - | 15 | 2081 | 6390688 | ||
BC060214.1 | - | 14 | 1961 | 6838657 |