Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprc6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153071.1 | NP_694711.1 | 3 | 922 | ||
GenBank mRNA | AY101365.1 | - | 3 | 922 | ||
AY735399.1 | - | 3 | 895 | |||
AY735401.1 | - | 3 | 895 | |||
BC108998.2 | - | 3 | 922 | 40048019 |
Set Membership: |
G-protein coupled receptor |