Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081278.2 | NP_001074747.2 | 2 | 1583 | ||
GenBank mRNA | AK031120.1 | - | 2 | 1427 | ||
AK039337.1 | - | 2 | 1584 | |||
AK044719.1 | - | 2 | 1225 | |||
BC064433.1 | - | 2 | 1207 | |||
BC080762.1 | - | 2 | 1201 | |||
BC117869.1 | - | 2 | 1059 | 40095526 | ||
BC117870.1 | - | 2 | 1059 | 40095531 |