Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncoa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111267.2 | NP_001104737.1 | 6 | 706 | ||
NM_172495.5 | NP_766083.3 | 16 | 3057 | |||
GenBank mRNA | AK033503.1 | - | 6 | 620 | ||
AK042369.1 | - | 5 | 991 | |||
AK128935.1 | - | 15 | 2880 | |||
AK165098.1 | - | 4 | 1394 | |||
AK171679.1 | - | 5 | 550 | |||
BC042791.1 | - | 1137 | ||||
BC049997.1 | - | 6 | 656 | |||
BC058572.1 | - | 6 | 628 | |||
BC076623.1 | - | 14 | 2663 | |||
BC080742.1 | - | 5 | 1812 | |||
BC141150.1 | - | 15 | 2758 | 9055762 | ||
BC145459.1 | - | 14 | 2643 | |||
EF602028.1 | - | 6 | 706 |