Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145378.4 | NP_663353.3 | 18, 19 | 2044 | ||
GenBank mRNA | AK037218.1 | - | 15, 16 | 3493 | ||
AK137721.1 | - | 17, 18 | 2273 | |||
BC042758.1 | - | 6, 7 | 923 | |||
DQ888308.1 | - | 17, 18 | 1952 |
Gene Symbol: Gm28042 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB195274.1 | - | 25, 26 | 3277 | ||
AB195275.1 | - | 24, 25 | 5385 | |||
BC098210.1 | - | 19, 20 | 2438 |