Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Szrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025608.2 | NP_001020779.1 | 3 | 186 | ||
NM_001277195.1 | NP_001264124.1 | 3 | 183 | |||
GenBank mRNA | AK027934.1 | - | 2 | 99 | ||
AK145912.1 | - | 2 | 106 | |||
AK171420.1 | - | 2 | 96 | |||
AK171702.1 | - | 2 | 111 | |||
BB871722.1 | - | 2 | 113 | |||
BC066992.1 | - | 3 | 134 | 5685985 | ||
CV784607.1 | - | 3 | 125 |