Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009331.3 | NP_033357.1 | 3 | 190 | ||
GenBank mRNA | AK037616.1 | - | 3 | 191 | ||
AK193329.1 | - | 1 | 36 | |||
BC056630.1 | - | 2 | 432 | |||
BC138596.1 | - | 2 | 170 | 8861616 | ||
BC145343.1 | - | 2 | 170 | 9053899 | ||
X61385.1 | - | 4 | 332 |