siRNA Details
Gene Symbol: Trim38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029935.2 | NP_001025106.1 | 1 | 254 | ||
GenBank mRNA | BC150836.1 | - | 1 | 254 | 9087747 | |
BC171998.1 | - | 1 | 254 |

Gene Symbol: Trim38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029935.2 | NP_001025106.1 | 1 | 254 | ||
GenBank mRNA | BC150836.1 | - | 1 | 254 | 9087747 | |
BC171998.1 | - | 1 | 254 |