siRNA Details
Gene Symbol: Ubtf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004961.1 | - | 16 | 1905 | ||
AK038139.1 | - | 12 | 3035 | |||
AK128947.1 | - | 16 | 1932 | |||
AK158861.1 | - | 16 | 1904 | |||
AK158935.1 | - | 17 | 1820 | |||
CA977198.1 | - | 2 | 226 | |||
X60831.1 | - | 16 | 1883 |
