Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TRP63 SIRNA Assays ›
          Gene Symbol:
          Trp63
          Gene Name:
          transformation related protein 63
          Gene Aliases:
          AI462811, Ket, P51/P63, P63, P73l, Tp63, Trp53rp1
          Chromosome Location:
          Chr.16: 25683763 - 25892103 on Build GRCm38
          UniGene ID:
          Mm.20894
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS278541
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Trp63
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001127259.1 NP_001120731.1 8 1592
          NM_001127260.1 NP_001120732.1 8 1592
          NM_001127261.1 NP_001120733.1 8 1592
          NM_001127262.1 NP_001120734.1 6 929
          NM_001127263.1 NP_001120735.1 6 929
          NM_001127264.1 NP_001120736.1 6 929
          NM_001127265.1 NP_001120737.1 6 929
          NM_011641.2 NP_035771.1 6 929
          GenBank mRNA AB010152.1 - 6 903
          AF075434.1 - 8 1067
          AF075435.1 - 8 1067
          AF075436.1 - 8 1067
          AF075437.1 - 6 785
          AF075438.1 - 6 785
          AF075439.1 - 6 785
          AK048623.1 - 6 929
          AK137253.1 - 6 930
          AK140023.1 - 2 182
          BC090649.1 - 6 911 30645427
          BC092537.1 - 6 929 30504012

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          P53-like transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          replicative cell aging
          skeletal system development
          establishment of planar polarity
          morphogenesis of a polarized epithelium
          hair follicle development
          positive regulation of mesenchymal cell proliferation
          epithelial cell development
          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          apoptotic process
          cellular response to DNA damage stimulus
          DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
          Notch signaling pathway
          multicellular organism development
          pattern specification process
          ectoderm and mesoderm interaction
          cell aging
          epidermis development
          organ morphogenesis
          proximal/distal pattern formation
          response to X-ray
          multicellular organism aging
          response to gamma radiation
          epidermal cell division
          regulation of epidermal cell division
          positive regulation of keratinocyte proliferation
          cell differentiation
          keratinocyte differentiation
          embryonic limb morphogenesis
          prostate gland development
          epithelial cell differentiation
          polarized epithelial cell differentiation
          hair follicle morphogenesis
          mitotic G1 DNA damage checkpoint
          cellular response to UV
          post-anal tail morphogenesis
          odontogenesis of dentin-containing tooth
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          negative regulation of apoptotic process
          regulation of cysteine-type endopeptidase activity involved in apoptotic process
          regulation of neuron apoptotic process
          skin morphogenesis
          keratinocyte proliferation
          negative regulation of keratinocyte differentiation
          positive regulation of osteoblast differentiation
          positive regulation of Notch signaling pathway
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          sympathetic nervous system development
          anatomical structure formation involved in morphogenesis
          smooth muscle tissue development
          female genitalia morphogenesis
          protein tetramerization
          protein homotetramerization
          neuron apoptotic process
          urinary bladder development
          cloacal septation
          prostatic bud formation
          squamous basal epithelial stem cell differentiation involved in prostate gland acinus development
          establishment of skin barrier
          positive regulation of cell cycle G1/S phase transition
          regulation of nucleic acid-templated transcription
          positive regulation of fibroblast apoptotic process
          negative regulation of mesoderm development
          negative regulation of cellular senescence
          positive regulation of apoptotic signaling pathway
          transcription factor activity, transcription factor binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          p53 binding
          DNA binding
          chromatin binding
          damaged DNA binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          protein domain specific binding
          identical protein binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          metal ion binding
          WW domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lns6n:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0