siRNA Details
Gene Symbol: Slmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144867.2 | NP_659116.1 | 4 | 410 | ||
GenBank mRNA | AK143220.1 | - | 4 | 394 | ||
BC019561.1 | - | 4 | 359 | 4459623 | ||
BY741062.1 | - | 4 | 411 |
