Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039149.1 | NP_001034238.1 | 3 | 343 | ||
NM_178687.2 | NP_848802.2 | 4 | 682 | |||
GenBank mRNA | AB195429.1 | - | 3 | 455 | ||
AF416980.1 | - | 4 | 514 | |||
AF421198.1 | - | 4 | 514 | |||
AF421199.1 | - | 3 | 175 | |||
AF421200.1 | - | 2 | 103 | |||
AK036455.1 | - | 5 | 777 | |||
AK140349.1 | - | 4 | 570 | |||
BC051526.1 | - | 3 | 277 | |||
BC132422.1 | - | 4 | 627 | 40130699 | ||
BC138769.1 | - | 4 | 627 | 8861791 | ||
BC145410.1 | - | 3 | 288 | 9053968 | ||
BC145411.1 | - | 4 | 627 |