Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033305.2 | NP_001028477.1 | 1 | 167 | ||
GenBank mRNA | AI853615.1 | - | 4 | 381 | ||
AK146762.1 | - | 1 | 169 | |||
BC147062.1 | - | 1 | 156 | 9056053 | ||
BC147063.1 | - | 1 | 156 | 9056059 |