Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skint6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103199.1 | NP_001096669.1 | 14 | 1342 | ||
GenBank mRNA | EU099304.1 | - | 14 | 1342 | ||
EU099305.1 | - | 14 | 1336 |