Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aim1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162970.1 | NP_001156442.1 | 20 | 4646 | ||
GenBank mRNA | AK145690.1 | - | 20 | 4653 | ||
AK155034.1 | - | 5 | 2147 | |||
AK165037.1 | - | 6 | 2607 | |||
BC042709.1 | - | 15 | 1667 | |||
BC077737.1 | - | 3 | 653 | |||
BP759371.1 | - | 1 | 159 |