Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172715.3 | NP_766303.1 | 5 | 800 | ||
GenBank mRNA | AK030171.1 | - | 6 | 985 | ||
AK138410.1 | - | 4 | 960 | |||
AK160261.1 | - | 5 | 745 | |||
BC096769.1 | - | 5 | 733 | 30101305 | ||
BC138228.1 | - | 4 | 553 | 8861247 | ||
BC145669.1 | - | 4 | 553 | 40130972 |