Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jazf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168277.1 | NP_001161749.1 | 4 | 587 | ||
NM_173406.3 | NP_775582.2 | 4 | 586 | |||
GenBank mRNA | AK043767.1 | - | 4 | 536 | ||
AK050474.1 | - | 4 | 558 | |||
BC048577.1 | - | 4 | 587 | 6706223 |