Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gnt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001036740.2 | NP_001031817.1 | 2 | 482 | ||
NM_146184.4 | NP_666296.1 | 1 | 874 | |||
GenBank mRNA | AK042388.1 | - | 1 | 857 | ||
AK154220.1 | - | 469 | ||||
AK154901.1 | - | 2 | 438 | |||
BC025206.1 | - | 2 | 468 | 5037295 |