Search
Search

Prc1| Gene Symbol: Prc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285997.1 | NP_001272926.1 | 12 | 1624 | ||
| NM_001285998.1 | NP_001272927.1 | 12 | 1633 | |||
| NM_145150.3 | NP_660132.2 | 12 | 1633 | |||
| GenBank mRNA | AF408435.1 | - | 4 | 392 | ||
| AK029140.1 | - | 11 | 2656 | |||
| AK088576.1 | - | 12 | 1605 | |||
| AK135778.1 | - | 10 | 1259 | |||
| BC005475.1 | - | 12 | 1558 | 3592138 | ||
| BC059808.1 | - | 12 | 1578 | |||