Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173767.3 | NP_776128.2 | 5 | 827 | ||
GenBank mRNA | AK014452.1 | - | 4 | 588 | ||
AK050143.1 | - | 2 | 322 | |||
BC026405.1 | - | 5 | 714 | 4211657 | ||
BC089586.1 | - | 2 | 331 | |||
BC139196.1 | - | 2 | 215 | 8862218 | ||
BC139197.1 | - | 2 | 215 | 8862219 | ||
DQ205645.1 | - | 4 | 540 |