Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spg7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153176.4 | NP_694816.3 | 14 | 1923 | ||
GenBank mRNA | AF512565.1 | - | 14 | 1955 | ||
AF547215.1 | - | 14 | 2252 | |||
AK018416.1 | - | 1 | 844 | |||
AK145540.1 | - | 14 | 1924 | |||
AK171647.1 | - | 11 | 2757 | |||
AK186971.1 | - | 3 | 282 | |||
AK196311.1 | - | 2 | 245 | |||
BC024466.1 | - | 8 | 984 | |||
BC024986.1 | - | 12 | 1578 | |||
BC051051.1 | - | 14 | 1857 | |||
BC055488.1 | - | 1768 | ||||
BC096690.1 | - | 7 | 823 | |||
BC138141.1 | - | 14 | 2150 | 8861160 |