Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Def8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253783.1 | NP_001240712.1 | 9 | 989 | ||
NM_001253784.1 | NP_001240713.1 | 8 | 967 | |||
NM_001281803.1 | NP_001268732.1 | 7 | 971 | |||
NM_054046.5 | NP_473387.1 | 7 | 880 | |||
GenBank mRNA | AK028726.1 | - | 8 | 939 | ||
AK034028.1 | - | 846 | ||||
AK046271.1 | - | 7 | 840 | |||
AK050405.1 | - | 8 | 938 | |||
AK081295.1 | - | 7 | 870 | |||
AK086174.1 | - | 7 | 842 | |||
AK087778.1 | - | 7 | 851 | |||
AK129019.1 | - | 7 | 922 | |||
AK133979.1 | - | 7 | 854 | |||
AK150334.1 | - | 7 | 816 | |||
AK150994.1 | - | 7 | 816 | |||
AK164147.1 | - | 7 | 852 | |||
AK182700.1 | - | 1 | 84 | |||
BC003306.1 | - | 9 | 940 | 3154712 | ||
CD545768.1 | - | 3 | 366 |