Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Def8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253783.1 | NP_001240712.1 | 10 | 1077 | ||
NM_001253784.1 | NP_001240713.1 | 9 | 1055 | |||
NM_001281803.1 | NP_001268732.1 | 8 | 1059 | |||
NM_054046.5 | NP_473387.1 | 8 | 968 | |||
GenBank mRNA | AK028726.1 | - | 9 | 1027 | ||
AK034028.1 | - | 934 | ||||
AK046271.1 | - | 8 | 928 | |||
AK050405.1 | - | 9 | 1026 | |||
AK081295.1 | - | 8 | 958 | |||
AK086174.1 | - | 8 | 931 | |||
AK087778.1 | - | 8 | 939 | |||
AK129019.1 | - | 8 | 1010 | |||
AK133979.1 | - | 8 | 942 | |||
AK150334.1 | - | 8 | 904 | |||
AK150994.1 | - | 8 | 904 | |||
AK164147.1 | - | 8 | 940 | |||
AK182700.1 | - | 2 | 172 | |||
BC003306.1 | - | 10 | 1028 | 3154712 | ||
CD545768.1 | - | 4 | 454 |