siRNA Details
Gene Symbol: Cdcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172873.3 | NP_766461.1 | 3 | 1521 | ||
GenBank mRNA | AK050720.1 | - | 3 | 1522 |

Gene Symbol: Cdcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172873.3 | NP_766461.1 | 3 | 1521 | ||
GenBank mRNA | AK050720.1 | - | 3 | 1522 |