Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081651.1 | NP_001075120.1 | 1 | 153 | ||
GenBank mRNA | AB232642.1 | - | 1 | 134 | ||
AK035722.1 | - | 1 | 70 | |||
BC132060.1 | - | 1 | 182 | 40130337 | ||
BC132062.1 | - | 1 | 182 | 40130339 |