Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153172.1 | NP_694812.1 | 6 | 867 | ||
GenBank mRNA | AF392062.1 | - | 6 | 867 | ||
AK077121.1 | - | 6 | 865 | |||
BC107230.1 | - | 5 | 840 | 40052042 | ||
BC107231.1 | - | 5 | 840 | 40052043 |