Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164089.1 | NP_001157561.1 | 4 | 453 | ||
NM_012003.2 | NP_036133.1 | 4 | 438 | |||
GenBank mRNA | AF071316.1 | - | 4 | 440 | ||
AK010417.1 | - | 4 | 448 | |||
AK010726.1 | - | 4 | 423 | |||
AK013155.1 | - | 4 | 406 | |||
BC003724.1 | - | 3 | 319 | 3593979 | ||
BG085353.2 | - | 4 | 438 |