Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mthfd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170785.1 | NP_001164256.1 | 16, 17 | 1893 | ||
NM_001170786.1 | NP_001164257.1 | 16, 17 | 1893 | |||
NM_172308.4 | NP_758512.3 | 16, 17 | 1893 | |||
GenBank mRNA | AK028211.1 | - | 16, 17 | 1844 | ||
AK036284.1 | - | 16, 17 | 1839 | |||
AK038579.1 | - | 16, 17 | 1851 | |||
AK075931.1 | - | 16, 17 | 1819 | |||
AK160025.1 | - | 16, 17 | 1825 | |||
BC030437.1 | - | 12, 13 | 1227 | 5374413 | ||
BC049936.1 | - | 12, 13 | 1216 | |||
BC058265.1 | - | 16, 17 | 1735 |