Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013759.2 | NP_038787.1 | 2 | 204 | ||
GenBank mRNA | AF195142.1 | - | 2 | 179 | ||
AK002339.1 | - | 2 | 164 | |||
AK002652.1 | - | 2 | 163 | |||
AK003112.1 | - | 2 | 164 | |||
AK003624.1 | - | 2 | 164 | |||
BC090646.1 | - | 2 | 204 | 30946285 | ||
BC141146.1 | - | 2 | 168 | 9055758 | ||
CX566836.1 | - | 2 | 204 |