Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013852.2 | NP_038880.1 | 14 | 1427 | ||
GenBank mRNA | AK030626.1 | - | 14 | 1392 | ||
AK181267.1 | - | 2 | 243 | |||
AK184016.1 | - | 2 | 243 | |||
AK198379.1 | - | 2 | 142 | |||
AK205676.1 | - | 2 | 143 | |||
BC032923.1 | - | 14 | 1368 | 1430389 |