Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013910.2 | NP_038938.1 | 8 | 1163 | ||
GenBank mRNA | AK031626.1 | - | 8 | 1129 | ||
AK034033.1 | - | 2 | 157 | |||
AK129479.1 | - | 19 | 2600 | |||
BC057622.1 | - | 8 | 1158 | 6402440 | ||
BC065090.1 | - | 19 | 2799 | 6851421 | ||
BC068239.1 | - | 16 | 2353 |